missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin A23, 99%, Thermo Scientific Chemicals

Product Code. 15493099
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493099 10 mg 10mg
15483099 5 mg 5mg
15403109 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493099 Supplier Thermo Scientific Alfa Aesar Supplier No. J60308.MA
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent inhibitor of EGF receptor kinase activity

Tyrphostin inhibit protein tyrosine kinase and block the EGF-dependent autophosphorylation of the receptor. The inhibitors can bind to the EGF receptor kinase domain with different affinities. It also inhibits PDGFR kinase. Tyrphostin A23 has been used to inhibit agonist-induced GP IIb- IIIa activation in permeabilized platelets and in other studies to block calcium channel currents in vascular smooth muscle cells. . Tyrphostin A23 can also inhibit the internalization of (125)I-transferrin in Heb7a cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrphostin inhibit protein tyrosine kinase and block the EGF-dependent autophosphorylation of the receptor. The inhibitors can bind to the EGF receptor kinase domain with different affinities. It also inhibits PDGFR kinase. Tyrphostin A23 has been used to inhibit agonist-induced GP IIb- IIIa activation in permeabilized platelets and in other studies to block calcium channel currents in vascular smooth muscle cells. . Tyrphostin A23 can also inhibit the internalization of (125)I-transferrin in Heb7a cells.

Solubility
Soluble in DMSO, ethanol, or DMF

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, acids, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118409-57-7
Molecular Formula C10H6N2O2
Molecular Weight (g/mol) 186.17
MDL Number MFCD00133899
InChI Key VTJXFTPMFYAJJU-UHFFFAOYSA-N
Synonym tyrphostin 23, tyrphostin a23, 3,4-dihydroxybenzylidene malononitrile, tyrphostin ag18, tyrphostin rg50810, unii-rv0gcd31oj, 2-3,4-dihydroxyphenyl methylidene propanedinitrile, 2-3,4-dihydroxybenzylidene malononitrile, alpha-cyano-3,4-dihydroxy cinnamonitrile, propanedinitrile,2-3,4-dihydroxyphenyl methylene
PubChem CID 2052
IUPAC Name 2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
SMILES C1=CC(=C(C=C1C=C(C#N)C#N)O)O

Specifications

Color Brown to Yellow
Quantity 10 mg
Solubility Information Soluble in DMSO, ethanol, or DMF
Formula Weight 186.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tyrphostin A23

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.