missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin 23, TRC

Product Code. 30449431
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449431 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449431 Supplier Toronto Research Chemicals Supplier No. TRCT923000250MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tyrphostin 23
CAS 118409-57-7
Molecular Formula C10 H6 N2 O2
Quantity 250 mg
InChI Formula InChI=1S/C10H6N2O2/c11-5-8(6-12)3-7-1-2-9(13)10(14)4-7/h1-4,13-14H
SMILES Oc1ccc(C=C(C#N)C#N)cc1O
IUPAC Name 2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
Molecular Weight (g/mol) 186.17
Formula Weight 186.0429
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.