missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin 23, MedChemExpress

Product Code. 16422797
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422797 50 mg 50mg
16406718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422797 Supplier Medchem Express Supplier No. HY15644
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tyrphostin 23 (Tyrphostin A23) is an EGFR inhibitor with an IC50 and Kiof 35 and 11 μM, respectively.

Tyrphostin 23 (Tyrphostin A23) is an EGFR inhibitor with an IC50 and Kiof 35 and 11 μM, respectively.

Specifications

Chemical Name or Material Tyrphostin 23
CAS 118409-57-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C10H6N2O2
Quantity 50 mg
Synonym Tyrphostin A23 RG-50810 AG 18
Solubility Information DMSO : 100 mg/mL (537.14 mM; Need ultrasonic)
SMILES N#C/C(C#N)=C/C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 186.17
Formula Weight 186.17
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.