missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYRP1 Antibody (TYRP1/3280), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79926IR
This item is not returnable.
View return policy
Description
TYRP1 Monoclonal specifically detects TYRP1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TYRP1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TYRP1 | |
| A recombinant fragment (around aa 257-377) of human TYRP1 protein (exact sequence is proprietary) (Uniprot: P17643) | |
| 0.1 mL | |
| Cancer | |
| This antibody reacts with It reacts with a 75kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TYRP1). | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| TYRP1/3280 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction