Learn More
TYRP1 Antibody (TYRP1/3280), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-79926V
Product Code. 18236347
  • £466.00 / 0.10mL
Description
TYRP1 Monoclonal specifically detects TYRP1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TYRP1
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
TYRP1
A recombinant fragment (around aa 257-377) of human TYRP1 protein (exact sequence is proprietary) (Uniprot: P17643)
0.1 mL
Cancer
This antibody reacts with It reacts with a 75kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TYRP1).
Store at 4°C in the dark.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TYRP1/3280
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP
Mouse
Protein A or G purified
RUO
Primary
Human
IgG2b κ