Learn More
Invitrogen™ Tyrosine Hydroxylase Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ P21962
Product Code. 11370672
  • £439.00 / 100µL
For Research Use Only. All usage must comply with product instructions.
Description
Description
Tyrosine Hydroxylase Polyclonal Antibody for Western Blot, IHC (F), ICC/IF
Tyrosine hydroxylase (TH) is an enzyme involved in the synthesis of catecholamine neurotransmitters dopamine, epinephrine, and norepinephrine. In all species, catecholamine synthesis is regulated by the interaction of TH with a cofactor, tetrahydrobiopterin (BH4). BH4 binds to the TH catalytic domain, resulting in enzymatic activity. Unlike TH in non-primate species, four human TH mRNA splice variants (hTH1-hTH4) have been isolated. These variants are identical in their catalytic domain, but differ in their N-terminal, regulatory domains. TH is also responsible for the conversion of L-tyrosine to L-dopa. TH plays a key role in the physiology of adrenergic neurons. The role of TH in the synthesis of catecholamine neurotransmitters suggests a correlation between the enzyme and a number of neuropathogenic diseases including: Parkinson's disease, schizophrenia, Segawa syndrome, and dystonia, as well as a variety of cardiovascular diseases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tyrosine Hydroxylase
Polyclonal
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
P04177, P07101
TH
SDS-denatured rat tyrosine hydroxylase, purified from pheochromocytoma.
100 μL
Primary
Human, Rat
Antibody
IgG
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Unconjugated
TH
dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase
Rabbit
Purified
RUO
25085, 7054
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Responsibly packaged