missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tyrosine Hydroxylase Polyclonal Antibody

Product Code. 11571982
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11571982 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11571982 Supplier Invitrogen™ Supplier No. PA118315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.

Tyrosine hydroxylase (TH) is an enzyme involved in the synthesis of catecholamine neurotransmitters dopamine, epinephrine, and norepinephrine. In all species, catecholamine synthesis is regulated by the interaction of TH with a cofactor, tetrahydrobiopterin (BH4). BH4 binds to the TH catalytic domain, resulting in enzymatic activity. Unlike TH in non-primate species, four human TH mRNA splice variants (hTH1-hTH4) have been isolated. These variants are identical in their catalytic domain, but differ in their N-terminal, regulatory domains. TH is also responsible for the conversion of L-tyrosine to L-dopa. TH plays a key role in the physiology of adrenergic neurons. The role of TH in the synthesis of catecholamine neurotransmitters suggests a correlation between the enzyme and a number of neuropathogenic diseases including: Parkinson's disease, schizophrenia, Segawa syndrome, and dystonia, as well as a variety of cardiovascular diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosine Hydroxylase
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with no preservative
Gene TH
Gene Accession No. P04177, P07101, P24529
Gene Alias dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase
Gene Symbols TH
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues P (63) R F I G R R Q S L I E D A R K(78) of human Tyrosine Hydroxylase conjugated to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100734150, 21823, 25085, 7054
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.