missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tyrosine Hydroxylase Monoclonal Antibody (4H2)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547336

 View more versions of this product

Product Code. 17805307

  • £1522.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Tyrosine Hydroxylase Monoclonal Antibody for Western Blot, IHC (PFA), ICC/IF

Tyrosine hydroxylase (TH) is an enzyme involved in the synthesis of catecholamine neurotransmitters dopamine, epinephrine, and norepinephrine. In all species, catecholamine synthesis is regulated by the interaction of TH with a cofactor, tetrahydrobiopterin (BH4). BH4 binds to the TH catalytic domain, resulting in enzymatic activity. Unlike TH in non-primate species, four human TH mRNA splice variants (hTH1-hTH4) have been isolated. These variants are identical in their catalytic domain, but differ in their N-terminal, regulatory domains. TH is also responsible for the conversion of L-tyrosine to L-dopa. TH plays a key role in the physiology of adrenergic neurons. The role of TH in the synthesis of catecholamine neurotransmitters suggests a correlation between the enzyme and a number of neuropathogenic diseases including: Parkinson's disease, schizophrenia, Segawa syndrome, and dystonia, as well as a variety of cardiovascular diseases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tyrosine Hydroxylase
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
P04177, P07101, P24529
TH
Full length human TH as expressed in and purified from E. coli.
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
4H2
Unconjugated
TH
dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase
Mouse
Protein G
RUO
21823, 25085, 7054
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.