missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosine Hydroxylase Antibody (OTI3H3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44650
This item is not returnable.
View return policy
Description
Tyrosine Hydroxylase Monoclonal antibody specifically detects Tyrosine Hydroxylase in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Tyrosine Hydroxylase | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI3H3 | |
| Western Blot 1:400∽4000, Immunocytochemistry/ Immunofluorescence 1:100 | |
| DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
| Full length human recombinant protein of human TH(NP_000351) produced in HEK293T cell. | |
| 100 μg | |
| Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer, Tyrosine Kinases | |
| 7054 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction