missing translation for 'onlineSavingsMsg'
Learn More

Tyrosine Hydroxylase Antibody (OTI1A4) [Alexa Fluor™ 647], Novus Biologicals™

Product Code. 30688298 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688298 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688298 Supplier Novus Biologicals Supplier No. NBP344864AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tyrosine Hydroxylase Monoclonal antibody specifically detects Tyrosine Hydroxylase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosine Hydroxylase
Applications Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone OTI1A4
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase
Host Species Mouse
Immunogen Full length human recombinant protein of human TH(NP_000351) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 7054
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.