Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 147 publications
Brand:  Novus Biologicals NB300-109
Product Code. 18757973
  • £498.00 / 0.10mL
Description
Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat, Drosophila, Insect, Mammal samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockout Validated, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1 mL