missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tyrosinase Recombinant Rabbit Monoclonal Antibody (6B2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA549843
This item is not returnable.
View return policy
Description
Tyrosinase Recombinant Monoclonal Antibody for ICC/IF, ELISA
The enzyme encoded by this gene catalyzes the first 2 steps, and at least 1 subsequent step, in the conversion of tyrosine to melanin. The enzyme has both tyrosine hydroxylase and dopa oxidase catalytic activities, and requires copper for function. Mutations in this gene result in oculocutaneous albinism, and nonpathologic polymorphisms result in skin pigmentation variation. The human genome contains a pseudogene similar to the 3' half of this gene.
Specifications
| Tyrosinase | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14679 | |
| TYR | |
| A synthesized peptide derived from human Tyrosinase. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 6B2 | |
| Unconjugated | |
| TYR | |
| albino; albino locus protein; ATN; C; CMM8; LB24-AB; monophenol monooxygenase; Oca1; OCA1A; OCAIA; oculocutaneous albinism IA; SHEP3; SK29-AB; skc35; Tumor rejection antigen AB; TYR; Tyrosinase; tyrosinase (oculocutaneous albinism IA); tyrosinase precursor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7299 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction