missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tyrosinase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599493
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Tyrosinase.
The enzyme encoded by this gene catalyzes the first 2 steps, and at least 1 subsequent step, in the conversion of tyrosine to melanin. The enzyme has both tyrosine hydroxylase and dopa oxidase catalytic activities, and requires copper for function. Mutations in this gene result in oculocutaneous albinism, and nonpathologic polymorphisms result in skin pigmentation variation. The human genome contains a pseudogene similar to the 3' half of this gene.
Specifications
| Tyrosinase | |
| Polyclonal | |
| Unconjugated | |
| TYR | |
| albino; albino locus protein; ATN; C; CMM8; LB24-AB; monophenol monooxygenase; Oca1; OCA1A; OCAIA; oculocutaneous albinism IA; SHEP3; SK29-AB; skc35; Tumor rejection antigen AB; TYR; Tyrosinase; tyrosinase (oculocutaneous albinism IA); tyrosinase precursor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22173, 308800, 7299 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11344, P14679 | |
| TYR | |
| A synthesized peptide derived from human TYR(Accession P14679), corresponding to amino acid residues A481-L529. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction