missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosinase Antibody (6B2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26264-50ul
This item is not returnable.
View return policy
Description
Tyrosinase Monoclonal antibody specifically detects Tyrosinase in Human samples. It is validated for ELISA, Immunocytochemistry
Specifications
| Tyrosinase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunocytochemistry | |
| 6B2 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| CMM8, EC 1.14.18.1, LB24-AB, Monophenol monooxygenase, OCA1A, OCAIA, SHEP3, SK29-AB, Tumor rejection antigen AB, tyrosinase, tyrosinase (oculocutaneous albinism IA) | |
| A synthesized peptide derived from Human Tyrosinase [UniProt P14679] | |
| 50 μL | |
| Lipid and Metabolism | |
| 7299 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction