missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TYRO3 Monoclonal Antibody (6D6F10)
GREENER_CHOICE

Product Code. 11375373
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375373 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11375373 Supplier Invitrogen™ Supplier No. MA515404
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-15404 targets Tyro3 in indirect ELISA, IF applications and shows reactivity with Human samples. The MA5-15404 immunogen is purified recombinant fragment of Tyro3 (aa138-321) expressed in E. Coli.

TYRO3 (tyrosine-protein kinase receptor TYRO3) is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including TULP1 or GAS6. It regulates many physiological processes including cell survival, migration, and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of TYRO3 on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by a ligand, TYRO3 plays a role in various processes such as neuron protection from excitotoxic injury, platelet aggregation, signal transduction, cell adhesion. and cytoskeleton reorganization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TYRO3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6D6F10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene TYRO3
Gene Accession No. P55146, Q06418
Gene Alias AI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; Dtk; Etk-2; etk2/tyro3; FLJ16467; Rek; Rse; Sky; soluble tyro 3; soluble tyro3; styro 3; styro3; TIF; TK19-2; tyro 3; TYRO3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase receptor TYRO3; Tyrosine-protein kinase RSE; tyrosine-protein kinase SKY; Tyrosine-protein kinase TIF
Gene Symbols TYRO3
Host Species Mouse
Immunogen Purified recombinant fragment of Tyro3 (aa138-321) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25232, 7301
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.