missing translation for 'onlineSavingsMsg'
Learn More

Tyramine, MedChemExpress

Product Code. 16463419
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16463419 Supplier Medchem Express Supplier No. HYW007606
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tyramine is an amino acid that helps regulate blood pressure. Tyramine occurs naturally in the body, and it's found in certain foods.

Tyramine is an amino acid that helps regulate blood pressure. Tyramine occurs naturally in the body, and it's found in certain foods.

Specifications

Chemical Name or Material Tyramine
CAS 51-67-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H11NO
Quantity 1 g
Solubility Information DMSO : 33.33 mg/mL (242.97 mM; Need ultrasonic) ∣H2O : 5.26 mg/mL (38.34 mM; Need ultrasonic)
SMILES C1=C(C=CC(=C1)O)CCN
Molecular Weight (g/mol) 137.18
Formula Weight 137.18
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.