missing translation for 'onlineSavingsMsg'
Learn More

Tyramine hydrochloride, 98%

Product Code. 11404903
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404903 100 g 100g
11484893 5 g 5g
11494893 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404903 Supplier Thermo Scientific Alfa Aesar Supplier No. A12220.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-19-5 | C8H12ClNO | 173.64 g/mol

Reagent for fluorescence enzyme immunoassay

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-19-5
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00012901
InChI Key RNISDHSYKZAWOK-UHFFFAOYSA-N
Synonym tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
PubChem CID 66449
IUPAC Name 4-(2-aminoethyl)phenol;hydrochloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C=C1

Specifications

Melting Point 271°C to 274°C
Quantity 100 g
Beilstein 3627058
Merck Index 14,9835
Formula Weight 173.65
Percent Purity 98%
Chemical Name or Material Tyramine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.