missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet UB-T-90

Product Code. 16397586 Shop All asecos Products
Change view
Click to view available options
Dimensions (D x W x H):
502 x 593 x 600 mm
600 x 593 x 800 mm
502 x 1102 x 600 mm
600 x 1102 x 800 mm
Door Style:
Left-hinged, Wing
Right-hinged, Wing
No. of Doors:
1
2
Includes:
(1) drawer max. interior height (sheet steel powder-coated (electrically conductive))
(1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated)
(1) perforated insert, (1) bottom collecting sump (stainless steel)
(1) shelf, (1) perforated insert (stainless steel), (1) bottom collecting sump (stainless steel)
(1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated)
(1) shelf, (1) perforated insert, (1) bottom collecting sump (stainless steel)
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Door Style No. of Doors Includes unitSize
16397586 600 x 593 x 800 mm Right-hinged, Wing 1 (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) Each
15938132 502 x 593 x 600 mm Left-hinged, Wing 1 (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) Each
15928132 502 x 1102 x 600 mm Left-hinged, Wing 2 (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) Each
16387576 600 x 593 x 800 mm Left-hinged, Wing 1 (1) drawer max. interior height (sheet steel powder-coated (electrically conductive)) Each
16317586 600 x 593 x 800 mm Left-hinged, Wing 1 (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) Each
16397576 600 x 593 x 800 mm Left-hinged, Wing 1 (1) perforated insert, (1) bottom collecting sump (stainless steel) Each
16327586 600 x 593 x 800 mm Left-hinged, Wing 1 (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) Each
16307586 600 x 593 x 800 mm Left-hinged, Wing 1 (1) shelf, (1) perforated insert (stainless steel), (1) bottom collecting sump (stainless steel) Each
16327596 600 x 1102 x 800 mm Left-hinged, Wing 2 (1) drawer max. interior height (sheet steel powder-coated (electrically conductive)) Each
16347596 600 x 1102 x 800 mm Left-hinged, Wing 2 (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) Each
16337596 600 x 1102 x 800 mm Left-hinged, Wing 2 (1) perforated insert, (1) bottom collecting sump (stainless steel) Each
15951342 600 x 1102 x 800 mm Left-hinged, Wing 2 (1) shelf, (1) perforated insert, (1) bottom collecting sump (stainless steel) Each
15985770 600 x 1102 x 800 mm Left-hinged, Wing 2 (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) Each
16357586 600 x 593 x 800 mm Right-hinged, Wing 1 (1) drawer max. interior height (sheet steel powder-coated (electrically conductive)) Each
16387586 600 x 593 x 800 mm Right-hinged, Wing 1 (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) Each
16367586 600 x 593 x 800 mm Right-hinged, Wing 1 (1) perforated insert, (1) bottom collecting sump (stainless steel) Each
16377586 600 x 593 x 800 mm Right-hinged, Wing 1 (1) shelf, (1) perforated insert (stainless steel), (1) bottom collecting sump (stainless steel) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy

For indoor use only

Product Code. 16397586 Supplier asecos™ Supplier No. 3591500133764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas

  • Robust construction and longevity: body made of powder-coated sheet steel
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Certificates

  • Fire resistance: 90 minutes
  • Type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
Depth (English) Interior 18.98 in.
Depth (Metric) Exterior 600 mm
Depth (Metric) Interior 482 mm
Dimensions (D x W x H) Interior 470 x 482 x 702 mm
Dimensions (D x W x H) 600 x 593 x 800 mm
Door Style Right-hinged, Wing
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (English) Interior 27.64 in.
Height (Metric) Exterior 800 mm
Height (Metric) Interior 702 mm
Includes (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated)
Load Bearing Capacity 30 kg (standard shelf)/75 kg (grid shelf)
Model UB90.080.059.060.TR
No. of Doors 1
No. of Shelves 1
Shelf Type Standard
Surface Coating Plain
Weight (English) 311 lb.
Weight (Metric) 141 kg
Width (English) Interior 18.50 in.
Width (Metric) Exterior 593 mm
Width (Metric) Interior 470 mm
Color Light Gray
Depth (English) Exterior 23.62 in.
Height (English) Exterior 31.50 in.
Material Gypsum
Product Type Safety Storage Cabinet (Under Bench)
Width (English) Exterior 23.35 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.