missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet

Product Code. 16377576 Shop All asecos Products
Change view
Click to view available options
Model:
UB90.060.059.TR
UB90.080.059.060.T
UB90.080.059.060.TR
UB90.080.110.060.2T
No. of Doors:
1
2
Shelf Type:
Shelf
Without Shelves
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model No. of Doors Shelf Type unitSize
16377576 UB90.080.059.060.T 1 Shelf Each
15914556 UB90.060.059.TR 1 Without Shelves Each
16307596 UB90.080.110.060.2T 2 Without Shelves Each
16317596 UB90.080.110.060.2T 2 Shelf Each
16337586 UB90.080.059.060.TR 1 Without Shelves Each
16347586 UB90.080.059.060.TR 1 Shelf Each
16367576 UB90.080.059.060.T 1 Without Shelves Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy

For indoor use only

Product Code. 16377576 Supplier asecos™ Supplier No. 3590000130486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For indoor use only

Safety storage under bench cabinet

  • Robust construction and longevity: body made of powder-coated sheet steel
  • Easy handling with comfort: door stops in any position, self-closing in the event of fire
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
  • Fire resistance 90 minutes
  • type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
Depth (Metric) Exterior 600 mm
Description Safety storage under bench cabinet UB-T-90 model UB90.080.059.060.T light gray (RAL 7035), interior equipment with 1 x shelf, 1 x perforated insert, 1 x bottom collecting sump (powder-coated sheet steel)
Dimensions (D x W x H) Interior 482 x 470 x 702 mm
Dimensions (D x W x H) 600 x 593 x 800 mm
Door Style Wing
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 800 mm
Includes Cabinet and interior equipment
Load Bearing Capacity 30 kg, 75 kg
Model UB90.080.059.060.T
No. of Doors 1
No. of Shelves 1
Shelf Type Shelf
Surface Coating Plain
Width (Metric) Exterior 593 mm
Width (Metric) 593 mm
Color Light Gray
Depth (English) Exterior 23.62 in.
Height (English) Exterior 31.5 in.
Height (Metric) Opening 702 mm
Material Gypsum
Product Type Safety Storage Cabinet (Under Bench)
Width (English) Exterior 23.35 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.