missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ S-Classic-90 Safety Storage Cabinet
For unrestricted storage of flammable hazardous materials in work areas.
Brand: Asecos™ 3020400230210
This item is not returnable.
View return policy
Description
- Type 90 safety storage cabinet
- With wing doors
- With cylinder locking
- Integrated adjusting aids to compensate uneven floor
- Fire resistance of 90 minutes
- In accordance with EN 14470-1 and TRGS 510
- GS-tested safety storage cabinets
Services
Five years
Specifications
EN 14470-1, DIN EN 14727, GS certified, CE compliant | |
616 mm | |
Body: Light grey RAL 7035, Door: Light grey RAL 7035; Sheet Steel RAL 7035 | |
Wing | |
1968 mm | |
S90.196.120 | |
Powder-coated | |
750 mm | |
Steel |
3 x shelf, 1 x bottom collecting sump (33L) | |
520 mm | |
524 x 1050 x 1742 mm | |
Increased protection against corrosion | |
1740 mm | |
3 | |
1196 mm | |
Gray | |
Safety Storage Cabinet |