missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet XL-CLASSIC-90

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3038200130384

Additional Details : Weight : 1,044.51843kg

 View more versions of this product

Product Code. 15944376

  • £6170.26 / Each
Estimated Shipment: 06-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

For indoor use only


Description

Description

  • User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
  • No unauthorised use: doors lockable with cylinder lock
  • Internal flexibility: transport base (optional) for safe and easy in-house transport
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Safety while decanting: end-to-end earthing link at the rear panel, inside with earthing wires incl. clamp
  • Certificates
    • Fire resistance 90 minutes
    • Type tested in accordance with EN 14470-1
    • GS certified
    • CE compliant
Specifications

Specifications

1890 mm
Safety storage cabinet
1386 mm
Powder-coated plain
1555 mm
1018 x 1555 x 2224 mm
864 x 1386 x 1890 mm
1018 mm
Wing
2
1 x transport base (steel powder-coated), 1 x bottom collecting sump with grid (sheet steel galvanized)
Safe and approved storage of flammable hazardous substances in working areas
864 mm
Steel
Door open arrest system
1147 kg
Light Gray
2224 mm
XL90.222.155.WDAS
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers