missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Safety Storage Cabinet V-MOVE-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos 3508505135100
Additional Details : Weight : 358.41599kg
Alert:
For indoor use only
Description
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
- Electronic opening and closing: the vertical drawers open and close at the push of a button
- Perfect access: the vertical drawers provide an overview and simple access to the stored containers
- Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
- No unauthorised use: vertical drawers lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
331 mm | |
77.40 in. | |
Gypsum | |
860 x 449 x 1966 mm | |
33.86 in. | |
331 x 775 x 1750 mm | |
807 lb. | |
4 | |
68.90 in. | |
Safety Storage Cabinet | |
17.68 in. | |
Silver, Anthracite Gray | |
1966 mm | |
V90.196.045.VDAC:0013 | |
1 |
775 mm | |
Textured | |
(4) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) | |
366 kg | |
30.51 in. | |
860 mm | |
50 kg | |
13.03 in. | |
1750 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
449 mm | |
Standard | |
Auto-close | |
300/285/285/380/290 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |