missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-PHOENIX-90 with 2 Doors

Product Code. 15376778 Shop All asecos Products
Change view
Click to view available options
Configuration:
3 Shelves, 1 Perforated Insert (370 mm), 1 Bottom Collecting Sump, 3 Tray Shelves, 1 Bottom Collecting Sump (PP), Vertical Dividing Wall
3 Shelves, 1 Perforated Insert (380 mm), 1 Bottom Collecting Sump
3 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump
3 Tray Shelves, 1 Perforated Insert (365 mm), 1 Bottom Collecting Sump
4 x drawer
6 Drawers, 3 Tray Shelves, 1 Bottom Collecting Sump, Vertical Dividing Wall
6 x drawer
Color:
Light Gray
Warning Yellow
Yellow
Width (Metric) Interior:
450 mm
1050 mm
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Color Width (Metric) Interior unitSize
15376778 3 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump Light Gray 1050 mm Each
15975946 3 Shelves, 1 Perforated Insert (370 mm), 1 Bottom Collecting Sump, 3 Tray Shelves, 1 Bottom Collecting Sump (PP), Vertical Dividing Wall Light Gray 450 mm Each
15912636 3 Shelves, 1 Perforated Insert (380 mm), 1 Bottom Collecting Sump Yellow 1050 mm Each
15912606 3 Tray Shelves, 1 Perforated Insert (365 mm), 1 Bottom Collecting Sump Light Gray 1050 mm Each
15952636 3 Tray Shelves, 1 Perforated Insert (365 mm), 1 Bottom Collecting Sump Warning Yellow 1050 mm Each
15962606 4 x drawer Light Gray 1050 mm Each
15902646 4 x drawer Yellow 1050 mm Each
15982606 6 x drawer Light Gray 1050 mm Each
15912646 6 x drawer Yellow 1050 mm Each
15985946 6 Drawers, 3 Tray Shelves, 1 Bottom Collecting Sump, Vertical Dividing Wall Light Gray 450 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy

For indoor use only

Product Code. 15376778 Supplier asecos™ Supplier No. 3011800130127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Space saving folding doors: less required space in front of the cabinet while doors are open
  • User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 3 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump
Depth (Metric) Exterior 616 mm
Depth (Metric) Interior 524 mm
Dimensions (D x W x H) Interior 524 x 1050 x 1742 mm
Dimensions (D x W x H) 616 x 1196 x 1968 mm
Door Style Folding
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1968 mm
Height (Metric) Interior 1742 mm
Includes Door open arrest system
Model S90.196.120.FDAS
No. of Doors 2
No. of Shelves 3
Shelf Type Standard
Surface Coating Powder-coated plain
Weight (Metric) 462.50 kg
Width (Metric) Exterior 1196 mm
Width (Metric) Interior 1050 mm
Color Light Gray
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.