missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90

Product Code. 15155155 Shop All asecos Products
Change view
Click to view available options
Configuration:
2 Shelves, 1 Perforated Insert (275 mm), 1 Bottom Collecting Sump
2 Shelves, 1 Perforated Insert (285 mm), 1 Bottom Collecting Sump
3 Shelves, 1 Perforated Insert (185 mm), 1 x 33 L Bottom Collecting Sump
3 Shelves, 1 Perforated Insert (185 mm), 1 x 33 L Bottom Collecting Sump
3 Shelves, 1 Perforated Insert (370 mm), 1 x 22 L Bottom Collecting Sump
3 Shelves, 1 Perforated Insert (380 mm), 1 33 L Bottom Collecting Sump
3 Tray Shelves, 1 Perforated Insert (111 mm), 1 Bottom Collecting Sump
3 Tray Shelves, 1 Perforated Insert (135 mm), 1 Bottom Collecting Sump
Color:
Light Gray
Warning Yellow
Yellow
Height (Metric) Exterior:
1298 mm
1968 mm
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Color Height (Metric) Exterior unitSize
15155155 2 Shelves, 1 Perforated Insert (275 mm), 1 Bottom Collecting Sump Yellow 1298 mm Each
15922786 2 Shelves, 1 Perforated Insert (285 mm), 1 Bottom Collecting Sump Light Gray 1298 mm Each
15566855 2 Shelves, 1 Perforated Insert (285 mm), 1 Bottom Collecting Sump Yellow 1298 mm Each
15932956 3 Shelves, 1 Perforated Insert (370 mm), 1 x 22 L Bottom Collecting Sump Light Gray 1968 mm Each
15902986 3 Shelves, 1 Perforated Insert (370 mm), 1 x 22 L Bottom Collecting Sump Yellow 1968 mm Each
15932786 3 Shelves, 1 Perforated Insert (185 mm), 1 x 33 L Bottom Collecting Sump Light Gray 1298 mm Each
15902816 3 Shelves, 1 Perforated Insert (185 mm), 1 x 33 L Bottom Collecting Sump Yellow 1298 mm Each
15536855 3 Shelves, 1 Perforated Insert (380 mm), 1 33 L Bottom Collecting Sump Warning Yellow 1968 mm Each
15317438 3 Tray Shelves, 1 Perforated Insert (111 mm), 1 Bottom Collecting Sump Light Gray 1968 mm Each
15992476 3 Tray Shelves, 1 Perforated Insert (135 mm), 1 Bottom Collecting Sump Light Gray 1968 mm Each
15992506 3 Tray Shelves, 1 Perforated Insert (135 mm), 1 Bottom Collecting Sump Warning Yellow 1968 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 15155155 Supplier asecos™ Supplier No. 3012100230155

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Safe and approved storage of flammable hazardous substances in working areas

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 2 Shelves, 1 Perforated Insert (275 mm), 1 Bottom Collecting Sump
Depth (Metric) Exterior 616 mm
Depth (Metric) Interior 524 mm
Dimensions (D x W x H) Interior 524 x 1050 x 1072 mm
Dimensions (D x W x H) 616 x 1196 x 1298 mm
Door Style Wing
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1298 mm
Height (Metric) Interior 1072 mm
Model S90.129.120
No. of Doors 2
No. of Shelves 3
Shelf Type Standard
Surface Coating Powder-coated plain
Weight (Metric) 337 kg
Width (Metric) Exterior 1196 mm
Width (Metric) Interior 1050 mm
Color Yellow
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.