missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System (Yellow)
Shop All asecos ProductsDescription
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Specifications
| Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
| Configuration | 3 Shelves, 1 Perforated Insert (380 mm), 1 Bottom Collecting Sump |
| Depth (Metric) Exterior | 616 mm |
| Depth (Metric) Interior | 524 mm |
| Dimensions (D x W x H) Interior | 524 x 1050 x 1742 mm |
| Dimensions (D x W x H) | 616 x 1196 x 1968 mm |
| Door Style | Wing |
| For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
| Height (Metric) Exterior | 1968 mm |
| Height (Metric) Interior | 1742 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?