missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System - Left-Hinged
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3022800230249
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
616 mm | |
524 x 450 x 1742 mm | |
Wing, Left-hinged | |
1968 mm | |
Door open arrest system | |
1 | |
Standard | |
281 kg | |
450 mm | |
Steel |
3 x shelf, 1 x perforated insert (335 mm), 1 x bottom collecting sump (sheet steel powder-coated) | |
524 mm | |
616 x 596 x 1968 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
S90.196.060.WDAS | |
3 | |
Powder-coated plain | |
596 mm | |
Yellow | |
Safety storage cabinet |