missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Safety Storage Cabinet S-CLASSIC-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos 30238-001-33741
Additional Details : Weight : 210.56850kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
450 mm | |
51.10 in. | |
Gypsum | |
616 x 596 x 1298 mm | |
24.25 in. | |
450 x 524 x 1072 mm | |
436 lb. | |
3 | |
42.20 in. | |
Safety Storage Cabinet | |
23.46 in. | |
Light Gray | |
1298 mm | |
S90.129.060 | |
1 |
524 mm | |
Plain | |
(3) drawers (sheet steel powder-coated) | |
198 kg | |
20.63 in. | |
616 mm | |
25 kg (standard shelf) | |
17.72 in. | |
1072 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
596 mm | |
Drawer | |
Wing | |
330/295/300 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |