missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Winged Doors

Product Code. 15997606 Shop All asecos Products
Change view
Click to view available options
Description:
Light gray doors
Warning Yellow doors
No. of Shelves:
3
6
Weight (Metric):
504.50 kg
528.50 kg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description No. of Shelves Weight (Metric) unitSize
15997606 Warning Yellow doors 3 528.50 kg Each
15967616 Light gray doors 3 528.50 kg Each
15987606 Warning Yellow doors 6 504.50 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy

For indoor use only

Product Code. 15997606 Supplier asecos™ Supplier No. 3421404033657

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas

  • Safe and approved storage of flammable hazardous substances in working areas
  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • EN 16121/16122 conformity
  • GS certified
  • CE compliant

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 6 x drawer (sheet steel powder-coated), 3 x tray shelf, 1 x bottom collecting sump (PP), vertical dividing wall
Depth (Metric) Exterior 615 mm
Depth (Metric) Interior 522 mm
Description Warning Yellow doors
Dimensions (D x W x H) Interior Left: 522 x 450 x 1647 mm, Right: 522 x 450 x 1647 mm
Dimensions (D x W x H) 615 x 1193 x 1953 mm
Door Style Wing
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1953 mm
Height (Metric) Interior 1647 mm
Model Q90.195.120.MV
No. of Doors 2
No. of Shelves 3
Shelf Type Tray shelf
Surface Coating Powder-coated textured
Weight (Metric) 528.50 kg
Width (Metric) Exterior 1193 mm
Width (Metric) Interior 450 mm
Color Anthracite Gray, Warning Yellow
Height (Metric) Opening 90 mm
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.