missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 Right-Hinged

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3100706134378

Additional Details : Weight : 199.37520kg

 View more versions of this product

Product Code. 15966766

  • £2250.38 / Each
Estimated Shipment: 12-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: door stops in any position, self-closing in the event of fire
  • No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
  • Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1626 mm
Safety storage cabinet
450 mm
Powder-coated textured
564 mm
620 x 564 x 1947 mm
519 x 450 x 1626 mm
620 mm
Wing, Right-hinged
90 mm
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
6 x drawer (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
519 mm
Steel
Entry height transport base
206.50 kg
Anthracite Gray, Light Gray
1947 mm
Q30.195.056.R
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers