missing translation for 'onlineSavingsMsg'
Learn More

Tylosin Tartrate Salt, MP BIOMEDICALS

Product Code. 11454802 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454802 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11454802 Supplier MP Biomedicals Supplier No. 0219345410
  • ONLINE EXCLUSIVE PRICE
    £264.22
    £226.80 / 10g
    Save £37.42
    14% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Application Notes:

Tylosin is commonly used in veterinary medicine and used to study:

  • protein synthesis at the level of the 50S ribosome
  • implant site abscess prevention in cattle
  • disposition kinetics and Mycoplasma gallisepticum and Mycoplasma meleagridis infections

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Antibiotic
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Powder
CAS 74610-55-2
Color Yellow
Quantity 10 g
Molecular Weight (g/mol) 1052.17
Chemical Name or Material Tylosin Tartrate Salt
compliance-icons compliance-icons
Product Identifier
  • tylosin tartarate salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: tylosin tartarate salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.