missing translation for 'onlineSavingsMsg'
Learn More

Tylosin 3-Acetate, TRC

Product Code. 17954035
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.5mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17954035 1 mg 1mg
17942125 0.5 mg 0.5mg
17943585 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17954035 Supplier Toronto Research Chemicals Supplier No. TRCT9476601MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63409-10-9
Molecular Formula C48H79NO18
Molecular Weight (g/mol) 958.14
Synonym Tylosin 3-acetate, Tylosin, 3-acetate, Oxacyclohexadecane, tylosin deriv., 3-O-Acetyltylosin, [4R-(4R*,5S*,6S*,7R*,9R*,11E,13E,15R*,16R*)]-4-Acetoxy-15-[[(6-deoxy-2,3-di-O-methyl-β-D-allopyranosyl)oxy]methyl]-6-[[3,6-dideoxy-4-O-(2,6-dideoxy-3-C-methyl-α-L-ribo-hexopyranosyl)-3-(dimethylamino)-β-D-glucopyranosyl]oxy]-16-ethyl-5,9,13-trimethyl-2,10-dioxooxacyclohexadeca-11,13-diene-7-acetaldehyde
IUPAC Name [(4R,5S,6S,7R,9R,11E,13E,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxymethyl]-5,9,13-trimethyl-2,10-dioxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-4-yl] acetate
SMILES CC[C@H]1OC(=O)C[C@@H](OC(=O)C)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(=C\[C@@H]1CO[C@@H]4O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]4OC)\C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C48H79NO18/c1-14-34-32(23-60-47-44(59-13)43(58-12)39(54)27(5)62-47)19-24(2)15-16-33(52)25(3)20-31(17-18-50)41(26(4)35(64-30(8)51)21-36(53)65-34)67-46-40(55)38(49(10)11)42(28(6)63-46)66-37-22-48(9,57)45(56)29(7)61-37/h15-16,18-19,25-29,31-32,34-35,37-47,54-57H,14,17,20-23H2,1-13H3/b16-15+,24-19+/t25-,26+,27-,28-,29+,31+,32-,34-,35-,37+,38-,39-,40-,41-,42-,43-,44-,45+,46+,47-,48-/m1/s1
Formula Weight 957.5297
Chemical Name or Material Tylosin 3-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.