missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TYK2, GST Tag Recombinant Protein
Tyrosine Kinase 2 (TYK2) is a member of the JAK kinase family
£436.00 - £11240.00
Specifications
Accession Number | NP_003322.2 |
---|---|
Gene Alias | TYK2 |
KinaseFamily | JAK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15314696
|
Invitrogen™
PR8440C |
100μg |
£1770.00
100µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468753
|
Invitrogen™
PV4791 |
1mg |
£11240.00
1mg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10546635
|
Invitrogen™
PV4790 |
10 μg |
£436.00
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tyrosine Kinase 2 (TYK2) plays both structural and catalytic roles in type I interferon (IFN) signaling.
Specifications
NP_003322.2 | |
JAK Kinase Family | |
Recombinant | |
Baculovirus |
TYK2 | |
Human | |
GST Tag |