Learn More
TYK2 Mouse anti-Human, Unconjugated, Clone: 1E1A10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67411-1-IG-150UL
 View more versions of this product
Product Code. 16830026
  • £349.00 / 150µL
Description
The TYK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TYK2. This antibody recognizes human antigen. The TYK2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TYK2
Monoclonal
800 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1E1A10
Unconjugated
150 μL
7297
IgG2b