missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Tygon™ LMT-55 Tubing without Configuration
Description
- Bio-based formulation delivers excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
- Resistant to flex fatigue and abrasion
- Less permeable than rubber tubing
- For use with virtually all inorganic chemicals found in the lab
Specifications
Specifications
| Description | 1.65 x 0.85 mm |
| Diameter (English) Inner | 0.06 in. |
| Diameter (Metric) Inner | 1.65 mm |
| Material | Tygon R3607 |
| Overall Length (English) | 32.8 ft. |
| Overall Length (Metric) | 10 m |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.85 mm |
| For Use With (Application) | For sample and reagent dispensing, biological and chemical assays, and water and soil analysis |
| Min. Operating Temperature (English) | -58°F |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?