missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ LMT-55 Tubing with Three Stop Configuration

Product Code. 11714648
Change view
Click to view available options
Diameter (Metric) Inner:
0.13 mm
0.51 mm
0.64 mm
0.76 mm
1.02 mm
1.14 mm
1.30 mm
1.42 mm
1.52 mm
1.65 mm
2.54 mm
2.79 mm
3.17 mm
Overall Length (Metric):
38.1 cm
10 m
Thickness (Metric):
0.85 mm
0.90 mm
Color:
Black/Black
Black/White
Blue/Blue
Crystal Clear
Gray/Gray
Orange/Orange
Orange/White
Orange/Yellow
Red/Red
Unit Size:
Pack of 12
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Overall Length (Metric) Thickness (Metric) Color unitSize
11714648 0.64 mm 38.1 cm 0.90 mm Orange/White Pack of 12
12340160 0.13 mm 38.1 cm 0.90 mm Orange/Orange Pack of 12
11704648 0.51 mm 38.1 cm 0.90 mm Orange/Yellow Pack of 12
10320881 0.76 mm 38.1 cm 0.85 mm Black/Black Pack of 12
15103216 1.14 mm 38.1 cm 0.85 mm Red/Red Pack of 12
11734648 1.30 mm 38.1 cm 0.85 mm Gray/Gray Pack of 12
15183206 1.65 mm 38.1 cm 0.85 mm Blue/Blue Pack of 12
11749744 3.17 mm 38.1 cm 0.85 mm Black/White Pack of 12
10773816 1.42 mm 38.1 cm 0.90 mm Crystal Clear Pack of 12
13474815 1.02 mm 10 m 0.85 mm Crystal Clear Pack of 12
15123216 2.54 mm 10 m 0.85 mm Crystal Clear Pack of 12
15133216 2.79 mm 10 m 0.85 mm Crystal Clear Pack of 12
15171773 1.52 mm 10 m 0.85 mm Crystal Clear Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 11714648 Supplier Saint Gobain Life Sciences Supplier No. 07053507ND
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ LMT-55 Tubing with Three Stop Configuration handles virtually all inorganic chemicals found in the lab.

  • Bio-based formulation delivers excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
TRUSTED_SUSTAINABILITY

Specifications

Color Code Orange/Orange
Diameter (English) Inner 0.03 in.
Diameter (Metric) Inner 0.64 mm
Material Tygon R3607
Overall Length (English) 1.25 ft.
Overall Length (Metric) 38.1 cm
Thickness (English) 0.04 in.
Thickness (Metric) 0.90 mm
For Use With (Application) For sample and reagent dispensing, biological and chemical assays, and water and soil analysis
Min. Operating Temperature (Metric) -50°C
Packaging Quantity 12 Pk.
Tubing Size 0.64 x 0.90 mm
Color Orange/White
Max. Operating Temperature (Metric) 74°C
Product Type Tubing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.