missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ismatec™ Tygon™ MHSL 2001 2-stop Tubing
Transparent, plasticizer-free 2-stop Tygon™ tubing with superior pump-life. Ismatec™ Tygon™ MHSL 2001 2-stop Tubing is especially designed for MEK and other aggressive solvents.
Brand: Ismatec™ SC0818
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Plasticizer and oil-free.
- Smooth inner-bore.
- Low sorption maintains fluid and tube integrity.
- Does not impart anything into the pumping medium.
- No release of hazardous materials when properly incinerated.
- Applications
- Acids: Excellent
- Alkaline solutions: Excellent
- Solvents: Good/Excellent
- Complies with: FDA.
- Sterilization
- Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C.
- Gas sterilization with ethylene oxide.
- Sterilization with radiation up to 2.5 mrad.
Order Info
DiameterMetricInside
Specifications
2-stop Tubing | |
Color-code: White-White | |
1.02 mm | |
2.72 mm | |
0.85 mm | |
For MEK and other aggressive solvents. CA cassettes | |
6 pcs./Pk. | |
1.02mm I.D., 2.72mm O.D. | |
Transparent |
FDA | |
Autoclavable | |
Tygon MHSL 2001 | |
38.1 cm | |
peristaltic pump | |
-73°C | |
Acids, Alkalies | |
CO2 1140, O2 76, N2 190 | |
57°C |