Learn More
DuPont™ Tychem™ 2000C Gown
Product Code. 17446683 Shop All DuPont Products
Change view
Click to view available options
Size:
Medium
2X-Large
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17446683 2X-Large Pack of 25
17475512 Medium Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17446683 Supplier DuPont™ Supplier No. D15546834
  • ONLINE EXCLUSIVE PRICE
    £518.42
    £403.16 / Pack of 25
    Save £115.26
    22% Off
View all promotions
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Shin-length gown with rear closure, knitted cuff with double cuff system, mandarin collar.
  • Double cuffs for increased protection and tighter connection with gloves
  • Elasticated wrists
  • Knitted cuff (inside) for better comfort
  • Offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure)
  • Used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Color Yellow
Disposable Single-use
Certifications/Compliance (EU) 2016/425, EN 14126, EN 1149-1