missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TY 52156

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TY 52156 is an S1P3 receptor antagonist (Ki = 110 nM); inhibits S1P-induced Ca2+ release in HUVEC cells. Suppresses FTY-720-induced S1P3 receptor-mediated bradycardia in vivo. Also inhibits S1P-induced breast cancer stem cell expansion in vitro. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | TY 52156 |
| Purity | >98% |
| Molecular Weight (g/mol) | 364.27 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction