missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNRD3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19517-1-AP-150UL
This item is not returnable.
View return policy
Description
Displays thioredoxin reductase, glutaredoxin and glutathione reductase activities. Catalyzes disulfide bond isomerization. Promotes disulfide bond formation between GPX4 and various sperm proteins and may play a role in sperm maturation by promoting formation of sperm structural components.Specifications
| TXNRD3 | |
| Polyclonal | |
| Unconjugated | |
| TXNRD3 | |
| TGR, thioredoxin reductase 3, Thioredoxin reductase TR2, TR2, TRXR3, TXNRD3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114112 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86VQ6 | |
| TXNRD3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction