missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNRD3 Polyclonal Antibody

TXNRD3 Polyclonal Antibody
Brand: Invitrogen PA565167
This item is not returnable.
View return policy
Description
TXNRD3 Polyclonal Antibody for ICC/IF
Specifications
| TXNRD3 | |
| Polyclonal | |
| Unconjugated | |
| TXNRD3 | |
| TGR; Thioredoxin and glutathione reductase; thioredoxin glutathione reductase; thioredoxin reductase 2; thioredoxin reductase 3; Thioredoxin reductase TR2; TR2; TRXR3; TXNRD3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86VQ6 | |
| TXNRD3 | |
| Recombinant Human TXNRD3. Recombinant protein control fragment (Product #RP-105917). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction