missing translation for 'onlineSavingsMsg'
Learn More

TXNRD2 Rabbit anti-Human, Mouse, Rat, Clone: 4O1K0, Novus Biologicals™

Product Code. 18343163
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343163 100 μg 100µL
18381447 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18343163 Supplier Bio-Techne Supplier No. NBP316715100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TXNRD2 Monoclonal antibody specifically detects TXNRD2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNRD2
Applications Western Blot
Classification Monoclonal
Clone 4O1K0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 1.8.1, EC 1.8.1.9, KIAA1652, Selenoprotein Z, SelZ, thioredoxin reductase 2, thioredoxin reductase 3, thioredoxin reductase beta, Thioredoxin reductase TR3, TR, TR3, TR-BETA, TRXR2thioredoxin reductase 2, mitochondrial
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TXNRD2 (Q9NNW7).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 10587
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.