missing translation for 'onlineSavingsMsg'
Learn More

TXNRD1 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

£119.00 - £295.00

Specifications

Antigen TXNRD1
Clone 1B10C4
Concentration 500 μg/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234127
View Documents
Proteintech
67728-1-IG-20UL
20 μL
ONLINE EXCLUSIVE PRICE
£135.00
£119.00 / 20µL
Save £16.00
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237516
View Documents
Proteintech
67728-1-IG-150UL
150 μL
ONLINE EXCLUSIVE PRICE
£349.00
£295.00 / 150µL
Save £54.00
15% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The TXNRD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TXNRD1. This antibody recognizes human, mouse, rat antigen. The TXNRD1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.
Specifications

Specifications

TXNRD1
500 μg/mL
Monoclonal
Liquid
RUO
7296
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
1B10C4
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Unconjugated
Mouse
Human, Mouse, Rat
GRIM-12; MGC9145; TR; TR1; TRXR1; TXNR
IgG1
Protein A purification
Monoclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.