missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNL4B Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | TXNL4B |
|---|---|
| Clone | 2 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18209397
|
Novus Biologicals
NBP2-90221-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286536
|
Novus Biologicals
NBP2-90221-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TXNL4B Monoclonal specifically detects TXNL4B in Human samples. It is validated for ELISA.Specifications
| TXNL4B | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Dim1-like protein, Dim2, DLPthioredoxin-like protein 4B, FLJ20511, thioredoxin-like 4B | |
| Recombinant Human TXNL4B protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 0.2 um filtered solution in PBS | |
| 54957 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title