missing translation for 'onlineSavingsMsg'
Learn More

TXNL4B Antibody (002), FITC, Novus Biologicals™

Product Code. 18383850 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383850 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383850 Supplier Novus Biologicals Supplier No. NBP290221F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TXNL4B Monoclonal specifically detects TXNL4B in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNL4B
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias Dim1-like protein, Dim2, DLPthioredoxin-like protein 4B, FLJ20511, thioredoxin-like 4B
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TXNL4B (Q9NX01; Met1-Ile149).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 54957
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.