missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXNL4A Polyclonal Antibody

Description
Immunogen sequence: MSYMLPHLHNG WQVDQAILSE EDRVVVIRFG HDWDPTCMKM DEVLYSIAEK VKNFAVIYLV DITEVPDFNK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Essential role in pre-mRNA splicing as component of the U5 snRNP and U4/U6-U5 tri-snRNP complexes that are involved in spliceosome assembly.
Specifications
Specifications
| Antigen | TXNL4A |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TXNL4A |
| Gene Accession No. | P83876 |
| Gene Alias | BMKS; D18Wsu98e; DIB1; Dim1; DIM1 protein homolog; ENSMUSG00000057130; SNRNP15; spliceosomal U5 snRNP-specific 15 kDa protein; thioredoxin like 4A; thioredoxin-like 4; thioredoxin-like 4A; thioredoxin-like protein 4A; Thioredoxin-like U5 snRNP protein U5-15kD; TXNL4; Txnl4a; U5-15kD; U5-15kDa |
| Gene Symbols | TXNL4A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?