missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNL4A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93215-0.1ml
This item is not returnable.
View return policy
Description
TXNL4A Polyclonal antibody specifically detects TXNL4A in Human, Mouse samples. It is validated for Western Blot
Specifications
| TXNL4A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DIB1, DIM1 protein homolog, DIM1Thioredoxin-like U5 snRNP protein U5-15kD, HsT161, Spliceosomal U5 snRNP-specific 15 kDa protein, thioredoxin-like 4, thioredoxin-like 4A, thioredoxin-like protein 4A, TXNL4, U5-15kD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-142 of human TXNL4A (NP_006692.1). MSYMLPHLHNGWQVDQAILSEEDRVVVIRFGHDWDPTCMKMDEVLYSIAEKVKNFAVIYLVDITEVPDFNKMYELYDPCTVMFFFRNKHIMIDLGTGNNNKINWAMEDKQEMVDIIETVYRGARKGRGLVVSPKDYSTKYRY | |
| 0.1 mL | |
| Apoptosis, Cell Biology | |
| 10907 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction