missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TXNL1 Polyclonal antibody specifically detects TXNL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Research Discipline | Apoptosis |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?