Learn More
Invitrogen™ TXNIP Recombinant Rabbit Monoclonal Antibody (JM60-35)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532771
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family.
Specifications
| TXNIP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q5M7W1, Q8BG60, Q9H3M7 | |
| TXNIP | |
| Synthetic peptide within Human TXNIP aa 50-99. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| JM60-35 | |
| Unconjugated | |
| TXNIP | |
| 1200008J08Rik; AA682105; ARRDC6; EST01027; HHCPA78; hyperlipidemia 1; Hyplip1; Tbp-2; THIF; thioredoxin binding protein 2; thioredoxin binding protein-2; thioredoxin interacting protein; thioredoxin-binding protein 2; Thioredoxin-interacting protein; TXNIP; upregulated by 1,25-dihydroxyvitamin D-3; VDUP1; Vitamin D3 up-regulated protein 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10628, 117514, 56338 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.