Learn More
TXNIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18243-1-AP-150UL
 View more versions of this product
Product Code. 16888304
  • £349.00 / 150µL
Description
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family.
Specifications
Specifications
TXNIP
Polyclonal
Unconjugated
TXNIP
EST01027, HHCPA78, THIF, Thioredoxin binding protein 2, TXNIP, VDUP1
Rabbit
Antigen Affinity Chromatography
RUO
10628, 56338
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BG60, Q9H3M7
Txnip
TXNIP Fusion Protein Ag13009
150 μL
Primary
Human, Mouse