missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TXNDC9 Recombinant Rabbit Monoclonal Antibody (K01_2J69)

Product Code. 30826576
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826576 50 μL
30824829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826576 Supplier Invitrogen™ Supplier No. MA565361

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TXNDC9 Recombinant Monoclonal Antibody for IHC (P), Western Blot

Not known. associated with cell differentiation. The protein encoded by this gene is a member of the thioredoxin family. The exact function of this protein is not known but it is associated with cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNDC9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2J69
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene TXNDC9
Gene Accession No. O14530
Gene Alias AI098020; APACD; ATP binding protein; ATP binding protein associated with cell differentiation; ATP-binding protein associated with cell differentiation; ES cell-related protein; fb15e07; PHLP3; Protein 1-4; thioredoxin domain containing 9; thioredoxin domain-containing protein 9; TXNDC9; wu:fb15e07; zgc:55522
Gene Symbols TXNDC9
Host Species Rabbit
Immunogen A synthetic peptide of human TXNDC9.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10190
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.